Learn More
Greiner Bio-One CELLSTAR μClear™ 96-well, Cell Culture-Treated, Flat-Bottom Microplate
Catalog No. 07001002
Change view
Click to view available options
Sterility:
Sterile
Catalog No. Sterility
07001002 Sterile
1 options
Catalog No. 07001002 Supplier Greiner Bio-One Supplier No. 65509099
Only null left
Color:Black, Clear Bottom; Sterility:Sterile; Lid:With Black Lid
  • Improved cell adhesion through physical surface treatment
  • Free of detectable DNase, RNase, human DNA, non-pyrogenic
  • Non-cytotoxic
  • Compatible with automated systems
  • Alphanumeric coding
  • High clarity; microplat
Specifications
Format Standard
Lid With Black Lid
Array 8 x 12
Packaging 8/Pk., 32/Cs.
Color Black, Clear Bottom
Material Polystyrene
No. of Wells 96
Sterility Sterile
Surface Treatment Cell Culture
Well Shape Flat Bottom (F-well)