Learn More
Greiner Bio-One CELLSTAR Cell-Repellent Surface Cell Culture Multiwell Plates
Ideal for suspension, spheroid cultures and stem cell aggregate formation. Greiner Bio-One CELLSTAR Cell-Repellent Surface Cell Culture Multiwell Plates effectively inhibit cell adhesion.
$265.60 - $289.10
Specifications
Bottom Shape Flat
Color Clear
Material Polystyrene
Sterility Sterile
Surface Treatment Cell Repellent
View More Specs
Products 3
Catalog Number Mfr. No. Well Count Price Quantity  
Catalog Number Mfr. No. Well Count Price Quantity  
07000651
missing translation for 'viewDocuments'
Greiner Bio-One
657970
6
Case of 5 Each for $265.60
Only null left
07000683
missing translation for 'viewDocuments' Promotion Details
Greiner Bio-One
662970
24
Case of 5 Each for $285.23
Only null left
07000728
missing translation for 'viewDocuments'
Greiner Bio-One
677970
48
Case of 5 Each for $289.10
Only null left
Description
Description
  • Made of polystyrene
  • Used for applications such as Gel-based 3D cultures and Magnetic 3D cell culturing (Nano3D technology)
  • Cell-repellent surface does not degrade or leach under common cell culture conditions, rendering an ideal substrate for native cell culture experiments
Service Plan
No
Warranty
No
Specifications
Specifications
Flat
Polystyrene
Cell Repellent
1
Clear
Sterile
5