Learn More
Alkali Scientific CellPro™ Transfer Pipette, 4mL Capacity, Short Stem, Non-Sterile, Bulk. 250/pk
Numéro de catalogue. 503027056
Encompass
These unbreakable, one-piece pipettes eliminate contaminated rubber bulbs, broken glass, and exposure to infectious materials. They are made of ultra-clear, low density polyethylene (LDPE) and are ideal for microbiology, blood banking, hematology, immunology, and urinalysis. Their low-affinity surface reduces sample, cell, or protein loss, and they can also be heat-sealed for airtight sample storage. The uniform drop size can be used for reproducible spotting of gels and TLC plates and dispensing into test plates, ID strips, PCR tubes, or 96-well plates. Sterile pipettes are also excellent for tissue culture application. Transparent LDPE Disposable & shatterproof; Heat sealable Precise, uniform drop size Nontoxic & inert Drops/mL: 22 & Bulb draw: 3.5mL Dimensions: 83mm L Temperature range: -196C to 75C
Numéro de catalogue. 503027056 Fournisseur Alkali Scientific Code fournisseur DP413200
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
These unbreakable, one-piece pipettes eliminate contaminated rubber bulbs, broken glass, and exposure to infectious materials. They are made of ultra-clear, low density polyethylene (LDPE) and are ideal for microbiology, blood banking, hematology, immunology, and urinalysis. Their low-affinity surface reduces sample, cell, or protein loss, and they can also be heat-sealed for airtight sample storage. The uniform drop size can be used for reproducible spotting of gels and TLC plates and dispensing into test plates, ID strips, PCR tubes, or 96-well plates. Sterile pipettes are also excellent for tissue culture application. Transparent LDPE Disposable & shatterproof; Heat sealable Precise, uniform drop size Nontoxic & inert Drops/mL: 22 & Bulb draw: 3.5mL Dimensions: 83mm L Temperature range: -196C to 75C