Learn More
Alkali Scientific CellPro™ Spectrophotometer Cuvette, PS, 10mm light path, 1000/Pk, 2.5mL semi-micro, round opening
Numéro de catalogue. 503028601
Encompass
Our disposable cuvettes are compatible with most popular spectrophotometers. Manufactured from high quality, optically clear polystyrene, they are intended for determinations in the 340-800nm (visible) wavelength range. Cuvettes are available in three sizes: 1.5mL micro, 2.5mL semi-micro and 4.5mL macro. All sizes are 12.5mm square, 45mm height and are compatible with various spectrophotometer models including: Carolina, Spectronic200, Spectronic20 and 20D, SpectronicEducator, Genesys™ and Accuris SmartDrop™. Fits most spectrophotometers and photometers Polystyrene for use in visible range (340-800nm) Protective packaging dust-proof and non-scratch Absorption reproducibility error of less than 1%**Caps are available and can be ordered separately.
Numéro de catalogue. 503028601 Fournisseur Alkali Scientific Code fournisseur EP4102
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Our disposable cuvettes are compatible with most popular spectrophotometers. Manufactured from high quality, optically clear polystyrene, they are intended for determinations in the 340-800nm (visible) wavelength range. Cuvettes are available in three sizes: 1.5mL micro, 2.5mL semi-micro and 4.5mL macro. All sizes are 12.5mm square, 45mm height and are compatible with various spectrophotometer models including: Carolina, Spectronic200, Spectronic20 and 20D, SpectronicEducator, Genesys™ and Accuris SmartDrop™. Fits most spectrophotometers and photometers Polystyrene for use in visible range (340-800nm) Protective packaging dust-proof and non-scratch Absorption reproducibility error of less than 1%**Caps are available and can be ordered separately.