Learn More
Alkali Scientific CellPro™ Sealing Mats for 96 Round Well Plates - 7.3mm, + open, 50/cs, DNase, RNase & Pyrogen-Free
Numéro de catalogue. 503026885
Encompass
Black alphanumeric coding Forms a tight seal in plate wells Perforated mats reseal themselves RNase, DNase and pyrogen free CellPro™ Sealing Mats are produced from high quality, chemically resistant silicone, and are designed to provide an airtight and liquid-tight seal. The material resists tearing and wear, minimizing the potential for particulate debris. They are compatible with temperatures from -40 to 80C, can be autoclaved and are also prepped for reuse with a 10% bleach wash with an ethanol rinse. They also feature black alphanumeric printing for easy reference and two types of resealable perforations (""+"" and ""-""), RNase, DNase and pyrogen free.
Numéro de catalogue. 503026885 Fournisseur Alkali Scientific Code fournisseur MPDS32013
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Black alphanumeric coding Forms a tight seal in plate wells Perforated mats reseal themselves RNase, DNase and pyrogen free CellPro™ Sealing Mats are produced from high quality, chemically resistant silicone, and are designed to provide an airtight and liquid-tight seal. The material resists tearing and wear, minimizing the potential for particulate debris. They are compatible with temperatures from -40 to 80C, can be autoclaved and are also prepped for reuse with a 10% bleach wash with an ethanol rinse. They also feature black alphanumeric printing for easy reference and two types of resealable perforations (""+"" and ""-""), RNase, DNase and pyrogen free.