Learn More
Alkali Scientific CellPro™ McCoy's 5A medium with L-glutamine, sodium bicarbonate, 500ml
Numéro de catalogue. 503028705
Encompass
Supports cell growth in research and biomanufacturing cell culture processes. Raw material components screened through strict quality control testing for lot-to-lot consistency. Product efficacy and homogeneity tested. Products are hydrated using purified process water and undergo sterile filtration.
Numéro de catalogue. 503028705 Fournisseur Alkali Scientific Code fournisseur MMC01
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Supports cell growth in research and biomanufacturing cell culture processes. Raw material components screened through strict quality control testing for lot-to-lot consistency. Product efficacy and homogeneity tested. Products are hydrated using purified process water and undergo sterile filtration.