Learn More
Alkali Scientific CellPro™ DMEM/F12 1:1 medium with L-glutamine, HEPES, 500ml. 10/Case
Numéro de catalogue. 503026806
Encompass
Supports cell growth in research and biomanufacturing cell culture processes. Raw material components screened through strict quality control testing for lot-to-lot consistency. Product efficacy and homogeneity tested. Products are hydrated using purified process water and undergo sterile filtration.
Numéro de catalogue. 503026806 Fournisseur Alkali Scientific Code fournisseur F12001CS
Supports cell growth in research and biomanufacturing cell culture processes. Raw material components screened through strict quality control testing for lot-to-lot consistency. Product efficacy and homogeneity tested. Products are hydrated using purified process water and undergo sterile filtration.