Learn More
Alkali Scientific CellPro™ DMEM, 1X, High Glucose, with L-glutamine, with Sodium Pyruvate, 500mL, 10/Case
Numéro de catalogue. 503027199
Encompass
Cellpro DMEM is unique from other media as it contains 4 times the concentration of amino acids and vitamins than the original Eagle's Minimal Essential Medium. DMEM was originally formulated with low glucose (1 g/L) and sodium pyruvate, but is often used with higher glucose levels, with or without sodium pyruvate. DMEM contains no proteins, lipids, or growth factors. Therefore, DMEM requires supplementation, commonly with 10%Fetal Bovine Serum(FBS). DMEM uses a sodium bicarbonate buffer system (3.7 g/L), and therefore requires a 510% CO2environment to maintain physiological pH.
Numéro de catalogue. 503027199 Fournisseur Alkali Scientific Code fournisseur DMEM13CS
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Cellpro DMEM is unique from other media as it contains 4 times the concentration of amino acids and vitamins than the original Eagle's Minimal Essential Medium. DMEM was originally formulated with low glucose (1 g/L) and sodium pyruvate, but is often used with higher glucose levels, with or without sodium pyruvate. DMEM contains no proteins, lipids, or growth factors. Therefore, DMEM requires supplementation, commonly with 10%Fetal Bovine Serum(FBS). DMEM uses a sodium bicarbonate buffer system (3.7 g/L), and therefore requires a 510% CO2environment to maintain physiological pH.