Learn More
Alkali Scientific CellPro™ 4mL Sterile Disposable Sampler Tubes, 1000/Cs, Separated Caps
Numéro de catalogue. 503029424
Encompass
Transport tubes are used in multiple sectors of the laboratory market, including medical, hospital, environmental, independent testing, food testing, and biological research labs. Used to store, transport or freeze samples. USP VI Polypropylene Tubes, HDPE Caps Self-standing: the vial placed on the lab workbench steadily even without a base Electron Beam Sterilization, SAL=10-6: safe, fast, environment-friendly, no chemical residues 100,000-level Clean Workshop Production, DNAse-free, RNAse-free, pyrogen-free, endotoxin-free Operating Temperature Range: Approx.-196C to 121C Excellent sealing performance, no leakage of negative pressure 95Kpa, ensuring safe and effective air transportation RNase, DNase, Pyrogen free
Numéro de catalogue. 503029424 Fournisseur Alkali Scientific Code fournisseur DST004SC
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Transport tubes are used in multiple sectors of the laboratory market, including medical, hospital, environmental, independent testing, food testing, and biological research labs. Used to store, transport or freeze samples. USP VI Polypropylene Tubes, HDPE Caps Self-standing: the vial placed on the lab workbench steadily even without a base Electron Beam Sterilization, SAL=10-6: safe, fast, environment-friendly, no chemical residues 100,000-level Clean Workshop Production, DNAse-free, RNAse-free, pyrogen-free, endotoxin-free Operating Temperature Range: Approx.-196C to 121C Excellent sealing performance, no leakage of negative pressure 95Kpa, ensuring safe and effective air transportation RNase, DNase, Pyrogen free