missing translation for 'onlineSavingsMsg'
Learn More

Cellotetraose 94.0+%, TCI America™

Catalog No. C279610MG
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C279610MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C279610MG Supplier TCI America Supplier No. C279610MG

Chemical Identifiers

CAS 38819-01-1
Molecular Formula C24H42O21
Molecular Weight (g/mol) 666.579
MDL Number MFCD00079079
InChI Key LUEWUZLMQUOBSB-HQXNUCTASA-N
Synonym cellotetraose
PubChem CID 131674892
IUPAC Name (2S,3S,5S)-2-[(3S,5S,6S)-6-[(3S,5S,6S)-4,5-dihydroxy-2-(hydroxymethyl)-6-[(3S,5S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-3-yl]oxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)OC3C(OC(C(C3O)O)OC4C(OC(C(C4O)O)O)CO)CO)CO)O)O)O)O

Specifications

Melting Point 253°C
Color White
Quantity 10 mg
Formula Weight 666.58
Percent Purity ≥94.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Cellotetraose
TSCA No
Recommended Storage Freezer