missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CellInsight™ CX7 High Content Analysis Platform

Catalog No. CX7A1110
Encompass
For research use only
Catalog No. CX7A1110 Supplier Thermo Scientific™ Supplier No. CX7A1110

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For research use only

Fast, automated cellular imaging and analysis platform designed for quantitative microscopy.

Integrated system for high content analysis

The Thermo Scientific™ CellInsight™ CX7 High Content Analysis (HCA) Platform is a fast, automated cellular imaging and analysis platform designed for quantitative microscopy.

The CellInsight CX7 HCA Platform offers an integrated way to develop and automate high content assays using a combination of 5-color brightfield and 7-color fluorescence imaging in confocal or wide-field modes. See how the CellInsight CX7 platform compares to other high content analysis platforms from Thermo Fisher Scientific.

Features of the CellInsight CX7 HCA Platform
  • 7-channel fluorescent imaging
  • 5-channel bright-field imaging
  • 7-channel confocal imaging
  • Software and laser-based autofocus for consistent scan times
  • Configured for fully automated plate handling and scanning

Capabilities
  • Addresses hundreds of assay types with flexible software and hardware tools
  • Scan and analyze microscope slides or most SBS-standard format microplates
  • Use HCS Studio software for integrated instrument control and data analysis
  • Achieve the fastest time to results by using intelligent acQuisition (iQ) real-time analysis feedback
  • Compatible with Molecular Probes™ fluorescent detection reagents
Hardware and Software
  • Stable and reliable light source enables reduced variability, increased data quality, and decreased operating costs
  • Broad reagent options with seven-color excitation from violet to near IR for assay multiplexing
  • Dual mode focusing using Software and Laser Autofocus methods to address both labelled and unlabeled sample types
  • Multi-mode imaging allows for wide-field, bright-field, and confocal imaging in one run
  • HCS Studio Cell Analysis Software is an integrated instrument control, visualization, and analysis software package for novice to advanced users
  • Scalable with Store Image and Database Software and optional Orbitor RS Microplate handler

Specifications

Product Line CellInsight
Description CellInsight™ CX7 High Content Analysis Platform
For Use With (Equipment) High Content Instrument
Color Assorted Colors
Detection Method Chromogenic, Fluorescence
Format Microplate, Slide(s)
Content And Storage • 1 Reader
Shipping Condition Room Temperature
Who do I contact if I have issues with my CellInsight systems or ArrayScan System?

Contact Instrument Customer Services for any issues regarding the CellInsight systems and ArrayScan System. They can be reached at instrumentservices@thermofisher.com.

For high-content analysis (HCA)/high-content screening (HCS), what are the main differences between the CellInsight CX5, CX7, and ArrayScan systems?

The basic differences are in the number of channels for excitation (5 versus 7), the ability to image in brightfield, widefield, and confocal, and the number of positions available on the objective turret. The CellInsight CX5 System is the most basic HCS system with illumination in 5 channels, imaging in brightfield and widefield, and a 1-position objective turret. The CellInsight CX7 System and ArrayScan System provide illumination in 7 channels, imaging in brightfield, widefield, and confocal, and either a 3-position (CX7) or 4-position objective turret (ArrayScan System). The ArrayScan System has the added option of live-cell imaging. For more information, go here (https://www.thermofisher.com/us/en/home/life-science/cell-analysis/cellular-imaging/high-content-screening.html).

For Research Use Only. Not for use in diagnostic procedures.