missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One CELLCOAT™ μClear™ 96-well, Poly-D Lysine-Treated, Flat-Bottom Microplate
Color:Black, Clear Bottom; Sterility:Non-sterile; Lid:With Lid
Supplier: Greiner Bio-One 655946
Description
- Reduced-serum or serum-free cultivation
- Cell differentiation and neuron growth
- Promotion of cell adhesion, proliferation and growth of transfected cell lines
- Increase in isolation and cultivation efficiency
Specifications
| Standard | |
| 8 x 12 | |
| Black, Clear Bottom | |
| 96 | |
| Non-sterile | |
| 392 μL | |
| 4800 x g (swinging bucket rotor) | |
| 5 | |
| Room Temperature |
| With Lid | |
| Not Autoclavable | |
| Polystyrene | |
| 25 to 340 μL | |
| Poly-D-Lysine | |
| Flat Bottom (F-well) | |
| 20 | |
| 20/Cs. | |
| Chimney |