Learn More
Perkin Elmer Canada UV Quartz SUPRASIL Rectangular Macro Cell with PTFE Stopper - 10 mm, Qty. 2
Catalog No. 509052053
Encompass
The UV Quartz SUPRASIL Macro Cell has a 10 mm path length, a cell volume of 3.5 mL, and a PTFE lid. Its outside dimensions are 45 mm (H) x 12.5 mm (W) x 12.5 mm (D). Qty. 2
Catalog No. 509052053 Supplier Perkin Elmer Canada Supplier No. B0631027
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The UV Quartz SUPRASIL Macro Cell has a 10 mm path length, a cell volume of 3.5 mL, and a PTFE lid. Its outside dimensions are 45 mm (H) x 12.5 mm (W) x 12.5 mm (D). Qty. 2