missing translation for 'onlineSavingsMsg'
Learn More

Celite 545, MilliporeSigma™

Catalog No. MCX05741
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MCX05741 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MCX05741 Supplier Technidata Supplier No. CX05741

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 68855-54-9
Molecular Formula CNa2O3
Molecular Weight (g/mol) 105.988
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
Synonym sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
PubChem CID 10340
ChEBI CHEBI:29377
IUPAC Name disodium;carbonate
SMILES C(=O)([O-])[O-].[Na+].[Na+]

Specifications

Physical Form Powder
Quantity 500 g
Linear Formula CaCl2
Formula Weight 11.98
Chemical Name or Material Celite™ 545
Recommended Storage Room Temperature