missing translation for 'onlineSavingsMsg'
Learn More

Cefotetan 98.0+%, TCI America™

Catalog No. C2936250MG
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2936250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C2936250MG Supplier TCI America Supplier No. C2936250MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 69712-56-7
Molecular Formula C17H17N7O8S4
Molecular Weight (g/mol) 575.604
MDL Number MFCD00864983
InChI Key SRZNHPXWXCNNDU-RHBCBLIFSA-N
Synonym cefotetan, cefotetanum, apacef, cefotetanum inn-latin, 6r,7s-7-4-2-amino-1-carboxy-2-oxoethylidene-1,3-dithietan-2-yl carbonyl amino-7-methoxy-3-1-methyl-1h-tetrazol-5-yl sulfanyl methyl-8-oxo-5-thia-1-azabicyclo 4.2.0 oct-2-ene-2-carboxylic acid, cefotan tn, 6r,7s-7-4-carbamoylcarboxymethylene-1,3-dithiethane-2-carboxamido-7-methoxy-3-1-methyl-1h-tetrazol-5-yl thio methyl-8-oxo-5-thia-1-azabicyclo 4.2.0 oct-2-ene-2-carboxylic acid, 7beta-4-2-amino-1-carboxy-2-oxoethylidene-1,3-dithietan-2-yl carbonyl amino-7alpha-methoxy-3-1-methyl-1h-tetrazol-5-yl sulfanyl methyl-3,4-didehydrocepham-4-carboxylic acid, 6r,7s-7-4-1-amino-3-hydroxy-1,3-dioxopropan-2-ylidene-1,3-dithietane-2-carbonyl amino-7-methoxy-3-1-methyltetrazol-5-yl sulfanylmethyl-8-oxo-5-thia-1-azabicyclo 4.2.0 oct-2-ene-2-carboxylic acid
PubChem CID 53025
IUPAC Name (6R,7S)-7-[[4-(2-amino-1-carboxy-2-oxoethylidene)-1,3-dithietane-2-carbonyl]amino]-7-methoxy-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
SMILES CN1C(=NN=N1)SCC2=C(N3C(C(C3=O)(NC(=O)C4SC(=C(C(=O)N)C(=O)O)S4)OC)SC2)C(=O)O

Specifications

Color White-Yellow
Quantity 250 mg
Formula Weight 575.60
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Cefotetan
RTECSNumber XI0330800
TSCA No
Recommended Storage Freezer (-20 °C)