missing translation for 'onlineSavingsMsg'
Learn More

Cefditoren Pivoxil 98.0+%, TCI America™

Catalog No. C2856200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2856200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C2856200MG Supplier TCI America Supplier No. C2856200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 117467-28-4
Molecular Formula C25H28N6O7S3
Molecular Weight (g/mol) 620.71
MDL Number MFCD00933166
InChI Key AFZFFLVORLEPPO-UHFFFAOYNA-N
PubChem CID 6437877
ChEBI CHEBI:560555
IUPAC Name {7-[2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetamido]-3-[2-(4-methyl-1,3-thiazol-5-yl)ethenyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carbonyloxy}methyl 2,2-dimethylpropanoate
SMILES CON=C(C(=O)NC1C2SCC(C=CC3=C(C)N=CS3)=C(N2C1=O)C(=O)OCOC(=O)C(C)(C)C)C1=CSC(N)=N1

Specifications

Melting Point 209°C
Color Yellow
Quantity 200 mg
Formula Weight 620.71
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Cefditoren Pivoxil
RTECSNumber XI0367800
TSCA No