missing translation for 'onlineSavingsMsg'
Learn More

Cefalotin Sodium, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™

Catalog No. 111022963
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022963 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022963 Supplier Merck Emd Millipore Supplier No. BP908

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

Specifications

Chemical Name or Material Pharmacopeia & Metrological Institutes Standards
CAS 58-71-9
Flash Point Not Applicable
Molecular Formula C16H15N2NaO6S2
Linear Formula C16H15N2NaO6S2
Quantity 100 mg
MDL Number MFCD00072025
Synonym Cephalothin sodium salt; 7-(2-Thienylacetamido)cephalosporanic acid sodium salt; Cephalotin sodium salt
Formula Weight 418.42
Grade BP