missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ CE Phosphoramidite, dG(iBu)

Catalog No. p-7083024
Encompass
Change view
Click to view available options
Quantity:
0.5 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
400331 0.5 g
400327 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 400331 Supplier Applied Biosystems™ Supplier No. 400331

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The dG(iBu) CE Phosphoramidite is used for nucleic acid synthesis on ABI DNA Synthesizers. Standard series of phosphoramidite deprotects in 6-8 hours at 55°C.β-cyanoethyl phosphoramidites are for use on ABI synthesizers to guarantee you high quality nucleic acid synthesis.

High QualityOur phosphoramidites benefit from proprietary manufacturing processes that yield high-purity products. In synthesis they perform at a correspondingly high level, producing pure, High Quality DNA at all scales. We guarantee that 5'-dimethoxytrityl nucleoside β-cyanoethyl phosphoramidites, monomer intermediates for licensed β-cyanoethyl phosphoramidite chemistry, will produce DNA that is at least 99.8% chemically authentic when used with recommended synthesis cycles.Dry phosphoroamidites (powder form) are guaranteed for one year from the date of shipment. Lifetime of diluted phosphoramidites in residence on the instrument is two weeks when used with CPG synthesis supports, and three weeks when used with polystyrene supports.
  • Proprietary manufacturing processes yield high-purity β-cyanoethyl phosphoramidites, which in turn guarantee production of high-quality DNA at all scales.
  • β-cyanoethyl phosphoramidites are use-tested and optimized for use on the ABI synthesizers to guarantee you high-quality nucleic acid synthesis.

Specifications

Chemical Name or Material dG(iBu) CEP
Content And Storage Store in freezer (-5 to -30°C).
Sample Type DNA
For Use With (Equipment) 3900 DNA Synthesizer
For Use With (Application) Oligo Synthesis
Nucleoside Type Deoxyguanosine (dG)
Nucleic Acid Type DNA
Product Type Oligonucleotide Synthesis Reagent
Quantity 0.5 g
Purity 99%
Reagent Type Phosphoramidite
Shipping Condition Room Temperature
Form Powder
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.