Learn More
SignalChem Biotech CDK9/CyclinT2, Active 50ug
Numéro de catalogue. 502575541
Encompass
Recombinant full-length human CDK9 and CyclinT2 proteins were co-expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Numéro de catalogue. 502575541 Fournisseur SignalChem Biotech Code fournisseur C4018BG50
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Recombinant full-length human CDK9 and CyclinT2 proteins were co-expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.