Learn More
SignalChem Biotech CDK1/CyclinA1, Active 500ug
Numéro de catalogue. 502575652
Encompass
Recombinant full-length human CDK1 and CyclinA1 were co-expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag on both proteins.
Numéro de catalogue. 502575652 Fournisseur SignalChem Biotech Code fournisseur C2218BG500
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Recombinant full-length human CDK1 and CyclinA1 were co-expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag on both proteins.