missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD65 Monoclonal Antibody (VIM8), FITC, eBioscience™, Invitrogen™
GREENER_CHOICE

Numéro de catalogue. 50245238
Modifier l'affichage
Click to view available options
Quantité:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
50245238 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 50245238 Fournisseur Invitrogen™ Code fournisseur 11065942

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Mouse Monoclonal Antibody

Description: This VIM8 antibody reacts with the desialylated asialoganglioside of human CD65. This type 2 fucosylated carbohydrate antigen is expressed on the surface of all myeloid cells during development. Moreover, CD65 is expressed highly on granulocytes and weakly on monocytes in peripheral blood. CD65 has been reported to be a ligand for CD62E (E-Selectin). However, the sialylated form of CD65 (designated CD65s) can be used as a marker for acute myeloid leukemia (AML) as its expression increases as CD34 decreases. Finally, CD65 is important for extravascular infiltration of AML cells. Applications Reported: This VIM8 antibody has been reported for use in flow cytometric analysis. Applications Tested: This VIM8 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.

This type 2 fucosylated carbohydrate antigen is expressed on the surface of all myeloid cells during development. Moreover, CD65 is expressed highly on granulocytes and weakly on monocytes in peripheral blood. CD65 has been reported to be a ligand for CD62E (E-Selectin). However, the sialylated form of CD65 (designated CD65s) can be used as a marker for acute myeloid leukemia (AML) as its expression increases as CD34 decreases. Finally, CD65 is important for extravascular infiltration of AML cells.

Spécifications

Antigène CD65
Applications Flow Cytometry
Classification Monoclonal
Clone VIM8
Concentration 5 μL/Test
Conjugué FITC
Formule PBS with BSA and 0.09% sodium azide; pH 7.2
Alias du gène Fucoganglioside Type 2; Fucoganglioside Type II
Espèces hôtes Mouse
Méthode de purification Affinity chromatography
Quantité 100 Tests
Statut réglementaire RUO
Primaire ou secondaire Primary
Espèces cibles Human
Contenu et stockage 4°C, store in dark, DO NOT FREEZE!
Type de produit Antibody
Forme Liquid
Isotype IgM
Afficher plus de résultats Afficher moins de résultats

For Research Use Only.