missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD53 Recombinant Mouse Monoclonal Antibody (161-2 (53/2))
GREENER_CHOICE

Numéro de catalogue. MA551887
Modifier l'affichage
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
MA551887 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. MA551887 Fournisseur Invitrogen™ Code fournisseur MA551887
Il en reste null

Mouse Recombinant Monoclonal Antibody

This antibody recognizes human CD53. It does not react with the baboon and horse antigen. Cd53 is a member of tetraspanin family and is involved in cellular activation as part of a signal transduction complex involving other membrane glycoproteins.

CD53 is a tetraspanin family transmembrane glycoprotein expressed in the lymphoid-myeloid lineage. This molecule has been reported to form complexes with other leukocyte surface proteins such as CD2, CD19, CD21, MHC II, VLA-4 or tetraspanins CD37, CD81 and CD82, thus probably modulating various signaling processes. CD53 is involved in radioresistancy of tumor cells and its triggering has anti-apoptotic effect. In thymus, CD53 is up-regulated in response to positive selection signals during T cell development, and is strongly expressed upon macrophage exposure to bacterial lipopolysaccharide, whereas stimulation of neutrophils results in down-regulation of CD53 expression.

Spécifications

Antigen CD53
Applications Flow Cytometry, Functional Assay, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 161-2 (53/2)
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene Cd53
Gene Accession No. P19397
Gene Alias AI323659; antigen MOX44 identified by monoclonal antibody MRC-OX44; Cd53; CD53 antigen; CD53 glycoprotein; CD53 molecule; CD53 tetraspan antigen; cell surface antigen; cell surface glycoprotein CD53; Leukocyte antigen MRC OX-44; leukocyte surface antigen CD53; MOX44; OX44; Ox-44; tetraspanin-25; transmembrane glycoprotein; TSPAN25; tspan-25
Gene Symbols Cd53
Host Species Mouse
Immunogen This antibody was raised by immunizing mice with stimulated human leukocytes.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 963
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Afficher plus de résultats Afficher moins de résultats