missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD52 Recombinant Mouse Monoclonal Antibody (CF1D12)

Mouse Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA551889
Description
This antibody binds to human CD52.
CD52, also known as CAMPATH-1 antigen, is a glycoprotein that is expressed on the surface of various immune cells, including T and B lymphocytes, natural killer cells, and monocytes. It is involved in the regulation of immune responses and is being studied as a potential therapeutic target in various diseases, including multiple sclerosis, chronic lymphocytic leukemia, and autoimmune disorders. CD52 is the target of alemtuzumab, a monoclonal antibody that is used to treat these conditions by depleting immune cells that express CD52.Identifiants chimiques
| CD52 | |
| 1 mg/mL | |
| Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 0.02% ProClin 300 | |
| P31358 | |
| 1043 | |
| Primary | |
| Protein A |
| CF1D12 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG3 κ | |
| Human | |
| CD52 | |
| AI463198; B7; B7-Ag; cambridge pathology 1 antigen; CAMPATH-1; CAMPATH-1 antigen; Cd52; CD52 antigen; CD52 antigen (CAMPATH-1 antigen); CD52 molecule; CDW52; CDW52 antigen (CAMPATH-1 antigen); CLS1; Epididymal secretory protein E5; epididymis secretory sperm binding protein Li 171mP; he5; HEL-S-171mP; human epididymis-specific protein 5; Lymphocyte differentiation antigen B7; Mb7 | |
| CD52 | |
| Antibody |
Spécifications
| CD52 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| P31358 | |
| CD52 | |
| Protein A | |
| RUO | |
| 1043 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry | |
| CF1D12 | |
| Unconjugated | |
| CD52 | |
| AI463198; B7; B7-Ag; cambridge pathology 1 antigen; CAMPATH-1; CAMPATH-1 antigen; Cd52; CD52 antigen; CD52 antigen (CAMPATH-1 antigen); CD52 molecule; CDW52; CDW52 antigen (CAMPATH-1 antigen); CLS1; Epididymal secretory protein E5; epididymis secretory sperm binding protein Li 171mP; he5; HEL-S-171mP; human epididymis-specific protein 5; Lymphocyte differentiation antigen B7; Mb7 | |
| Mouse | |
| 200 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG3 κ |