missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD51 Polyclonal Antibody

Rabbit Polyclonal Antibody
$286.18 - $821.34
Spécifications
| Antigen | CD51 |
|---|---|
| Concentration | 0.5 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantity & Availability | |||||
PIPA5119210
![]() |
Invitrogen™
PA5119210 |
200 μL |
chaque for $821.34
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
PIPA5122108
|
Invitrogen™
PA5122108 |
20 μL |
chaque for $286.18
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
CD51 Polyclonal Antibody for Western Blot, ICC/IF
ITAGV encodes integrin alpha chain V. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. The I-domain containing integrin alpha V undergoes post-translational cleavage to yield disulfide-linked heavy and light chains, that combine with multiple integrin beta chains to form different integrins. Among the known associating beta chains (beta chains 1,3,5,6, and 8, 'ITGB1', 'ITGB3', 'ITGB5', 'ITGB6', and 'ITGB8'), each can interact with extracellular matrix ligands, the alpha V beta 3 integrin, perhaps the most studied of these, is referred to as the Vitronectin receptor (VNR). In addition to adhesion, many integrins are known to facilitate signal transduction. The ITGB3 protein product is the integrin beta chain beta 3. Integrins are integral cell-surface proteins composed of an alpha chain and a beta chain. A given chain may combine with multiple partners resulting in different integrins. Integrin beta 3 is found along with the alpha IIb chain in platelets. Integrins are known to participate in cell adhesion as well as cell-surface mediated signalling.Spécifications
| CD51 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human | |
| ITGAV | |
| 1110004F14Rik; 2610028E01Rik; alpha v integrin; antigen identified by monoclonal antibody L230; Cd51; CD61; D430040G12Rik; Dnmt3l-ps1 pseudogene; GP3A; GPIIIa; integrin alpha V; integrin alpha-V; Integrin alpha-V heavy chain; Integrin alpha-V light chain; integrin alphaVbeta3; integrin subunit alpha V; integrin, alpha V; integrin, alpha V (vitronectin receptor, alpha polypeptide, antigen CD51); ITGAV; ITGB3; MSK8; Vitronectin receptor; vitronectin receptor alpha polypeptide (VNRA); vitronectin receptor subunit alpha; VNR; VNRA; VTNR | |
| ITGAV | |
| Primary | |
| Antigen affinity chromatography, Protein A |
| 0.5 mg/mL | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P06756 | |
| 3685 | |
| Recombinant protein Integrin Alpha V. The antigen corresponds to amino acid range 560-744 of the target protein. | |
| Antibody |