Learn More
ACROBiosystems Inc Recombinant Protein;Human CD47 Protein, Fc Tag (HPLC-verified);HEK293;1MG;CD7,CD47,MER6,IAP,OA3
Numéro de catalogue. 501658082
Encompass
Recombinant Protein;Human CD47 Protein, Fc Tag (HPLC-verified);1MG;Human CD47, Fc Tag (CD7-H5256) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Pro 139 (Accession # NP_942088).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 40.4 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD47, Fc Tag (HPLC-verified) (Cat. No. CD7-H5256) at 2 g/mL can bind Human SIRP alpha, His Tag (Cat. No. SIA-H5225) with a linear range of 16-250 ng/mL (QC tested).
Numéro de catalogue. 501658082 Fournisseur ACROBiosystems Inc Code fournisseur CD7H52561MG
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Recombinant Protein;Human CD47 Protein, Fc Tag (HPLC-verified);1MG;Human CD47, Fc Tag (CD7-H5256) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Pro 139 (Accession # NP_942088).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 40.4 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD47, Fc Tag (HPLC-verified) (Cat. No. CD7-H5256) at 2 g/mL can bind Human SIRP alpha, His Tag (Cat. No. SIA-H5225) with a linear range of 16-250 ng/mL (QC tested).