Learn More
ACROBiosystems Inc ACROBiosystems Biotinylated Mouse CD47 Protein, His,Avitag™
Numéro de catalogue. 501956688
Encompass
Recombinant Protein;25UG;Biotinylated Mouse CD47, His,Avitag (CD7-M82E4) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Lys 140 (Accession # Q61735-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 17.4 kDa. The protein migrates as 30-48 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Mouse CD47, His,Avitag (Cat. No. CD7-M82E4) at 2 g/mL (100 L/well) on streptavidin precoated (0.5 g/well) plate, can bind Mouse SIRP alpha, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. SIA-M5252) with a linear range of 0.019-0.625 g/mL (QC tested).
Numéro de catalogue. 501956688 Fournisseur ACROBiosystems Inc Code fournisseur CD7M82E425UG
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Recombinant Protein;25UG;Biotinylated Mouse CD47, His,Avitag (CD7-M82E4) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Lys 140 (Accession # Q61735-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 17.4 kDa. The protein migrates as 30-48 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Mouse CD47, His,Avitag (Cat. No. CD7-M82E4) at 2 g/mL (100 L/well) on streptavidin precoated (0.5 g/well) plate, can bind Mouse SIRP alpha, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. SIA-M5252) with a linear range of 0.019-0.625 g/mL (QC tested).