Learn More
ACROBiosystems Inc ACROBiosystems Rat CD47 Protein, Fc Tag
Numéro de catalogue. 501957369
Encompass
Recombinant Protein;1MG;Rat CD47, Fc Tag (CD7-R5256) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Lys 140 (Accession # P97829).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 40.4 kDa. As a result of glycosylation, the protein migrates as 50-66 kDa under reducing (R) condition, and 100-130 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Mouse SIRP alpha, Mouse IgG2a Fc Tag (Cat. No. SIA-M5252) at 10 g/mL (100 L/well) can bind Rat CD47, Fc Tag (Cat. No. CD7-R5256) with a linear range of 2-20 ng/mL (QC tested).
Numéro de catalogue. 501957369 Fournisseur ACROBiosystems Inc Code fournisseur CD7R52561MG
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Recombinant Protein;1MG;Rat CD47, Fc Tag (CD7-R5256) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Lys 140 (Accession # P97829).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 40.4 kDa. As a result of glycosylation, the protein migrates as 50-66 kDa under reducing (R) condition, and 100-130 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Mouse SIRP alpha, Mouse IgG2a Fc Tag (Cat. No. SIA-M5252) at 10 g/mL (100 L/well) can bind Rat CD47, Fc Tag (Cat. No. CD7-R5256) with a linear range of 2-20 ng/mL (QC tested).