missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD45.2 Monoclonal Antibody (104), PE-Cyanine7, eBioscience™, Invitrogen™

Numéro de catalogue. 501123292
Modifier l'affichage
Click to view available options
Quantité:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
501123292 25 μg
501123293 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 501123292 Fournisseur Invitrogen™ Code fournisseur 25045480
Saison des économies
Afficher toutes les promotions

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Mouse Monoclonal Antibody

Description: The 104 monoclonal antibody reacts with the mouse CD45 molecule, the leukocyte common antigen (LCA) in CD45.2-expressing mouse strains. The strains that express CD45.2 include the most commonly used mouse strains C57BL/6, BALB/c, C58, DBA/1, DBA/2, C3H/He, CBA, 129, A and AKR. CD45.2 is expressed by all leukocytes in these strains. Applications Reported: This 104 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 104 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation.

CD45.2 (LCA, leukocyte common antigen) is a receptor-type protein tyrosine phosphatase ubiquitously expressed in all nucleated hematopoietic cells, comprising approximately 10% of all surface proteins in lymphocytes. CD45.2 glycoprotein is crucial in lymphocyte development and antigen signaling, serving as an important regulator of Src-family kinases. CD45.2 protein exists as multiple isoforms as a result of alternative splicing; these isoforms differ in their extracellular domains, whereas they share identical transmembrane and cytoplasmic domains. These isoforms differ in their ability to translocate into the glycosphingolipid-enriched membrane domains and their expression depends on cell type and physiological state of the cell. Besides the role in immunoreceptor signaling, CD45.2 is important in promoting cell survival by modulating integrin-mediated signal transduction pathway and is also involved in DNA fragmentation during apoptosis. CD45RA is an isoform of the CD45 complex and has restricted expression between different subtypes of lymphoid cells.

Spécifications

Antigène CD45.2
Applications Flow Cytometry
Classification Monoclonal
Clone 104
Concentration 0.2 mg/mL
Conjugué PE-Cyanine7
Formule PBS with 0.09% sodium azide; pH 7.2
Gène Ptprc
Numéro d’acquisition du gène P06800
Alias du gène B220; Cd45; CD45 antigen; CD45R; GP180; LCA; L-CA; leukocyte common antigen; loc; LY5; Ly-5; lymphocyte antigen 5; lymphocyte common antigen; Lyt-4; protein tyrosine phosphatase, receptor type, C; Ptprc; Receptor-type tyrosine-protein phosphatase C; T200; T200 glycoprotein
Symboles de gène Ptprc
Espèces hôtes Mouse
Méthode de purification Affinity chromatography
Quantité 25 μg
Statut réglementaire RUO
Primaire ou secondaire Primary
ID du gène (Entrez) 19264
Espèces cibles Mouse
Contenu et stockage 4°C, store in dark, DO NOT FREEZE!
Type de produit Antibody
Forme Liquid
Isotype IgG2a κ
Afficher plus de résultats Afficher moins de résultats

For Research Use Only.