Learn More
ACROBiosystems Inc Recombinant Protein;Human CD40 / TNFRSF5 Protein, Fc Tag (HPLC-verified);HEK293;1MG;CD0,CD40,CD-40,Bp50,Bp-50,CDW40,MGC9013,TNFRSF5,TNFRSF-5,p50,p-50
Numéro de catalogue. 501657496
Encompass
Recombinant Protein;Human CD40 / TNFRSF5 Protein, Fc Tag (HPLC-verified);1MG;Human CD40, Fc Tag (CD0-H5253) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Arg 193 (Accession # NP_001241.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 45.3 kDa. The protein migrates as 50-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD40 Ligand, His Tag (Cat. No. CDL-H5248) at 5 g/mL (100 L/well) can bind Human CD40, Fc Tag (HPLC-verified) (Cat. No. CD0-H5253) with a linear range of 0.039-1.25 g/mL (QC tested).;FACS assay shows that recombinant Human CD40, Fc Tag (HPLC-verified) (Cat. No. CD0-H5253) can bind to 293T cells overexpressing human CD40L, and the binding is inhibited by neutralizing Anti-Human CD40 MAb. The concentration of CD40 and Anti-Human CD40 MAb used are 0.03 g/ml and 0.3 g/ml respectively.
Numéro de catalogue. 501657496 Fournisseur ACROBiosystems Inc Code fournisseur CD0H52531MG
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Recombinant Protein;Human CD40 / TNFRSF5 Protein, Fc Tag (HPLC-verified);1MG;Human CD40, Fc Tag (CD0-H5253) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Arg 193 (Accession # NP_001241.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 45.3 kDa. The protein migrates as 50-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD40 Ligand, His Tag (Cat. No. CDL-H5248) at 5 g/mL (100 L/well) can bind Human CD40, Fc Tag (HPLC-verified) (Cat. No. CD0-H5253) with a linear range of 0.039-1.25 g/mL (QC tested).;FACS assay shows that recombinant Human CD40, Fc Tag (HPLC-verified) (Cat. No. CD0-H5253) can bind to 293T cells overexpressing human CD40L, and the binding is inhibited by neutralizing Anti-Human CD40 MAb. The concentration of CD40 and Anti-Human CD40 MAb used are 0.03 g/ml and 0.3 g/ml respectively.