missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD290 (TLR10) Monoclonal Antibody (3C10C5), PE, eBioscience™
GREENER_CHOICE

Numéro de catalogue. 12290942
Encompass
Modifier l'affichage
Click to view available options
Quantité:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
12290942 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 12290942 Fournisseur Invitrogen™ Code fournisseur 12290942
Saison des économies
Afficher toutes les promotions

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Mouse Monoclonal Antibody

Description: This 3C10C5 monoclonal antibody reacts with human CD290, which is also known as Toll-like receptor 10 (TLR10). This type I transmembrane glycoprotein is expressed on B cells, monocytes, regulatory T cells (Tregs), and plasmacytoid dendritic cells. Moreover, CD290 expression is dependent on Foxp3 and NFAT in Tregs. Studies demonstrate that CD290 can self-dimerize, as well as heterodimerize with other members of the TLR family such as TLR1 and TLR2. Ligand binding results in association of the receptor with the adapter molecule MyD88 and subsequent activation of signaling pathways. Although the ligands of CD290 remain undefined, this receptor has been associated with the immune response against microbial and fungal infection and autoimmune diseases such as Crohn's disease and systemic lupus erythematosous. Applications Reported: This 3C10C5 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 3C10C5 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: μ8 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.

The protein encoded by this gene is a member of the Toll-like receptor (TLR) family which plays a fundamental role in pathogen recognition and activation of innate immunity. TLRs are highly conserved from Drosophila to humans and share structural and functional similarities. They recognize pathogen-associated molecular patterns (PAMPs) that are expressed on infectious agents, and mediate the production of cytokines necessary for the development of effective immunity. The various TLRs exhibit different patterns of expression. This gene is most highly expressed in lymphoid tissues such as spleen, lymph node, thymus, and tonsil. Its exact function is not known. Multiple alternatively spliced transcript variants encoding the same protein have been found for this gene.

Spécifications

Antigène CD290 (TLR10)
Applications Flow Cytometry
Classification Monoclonal
Clone 3C10C5
Concentration 5 μL/Test
Conjugué PE
Formule PBS with BSA and 0.09% sodium azide; pH 7.2
Gène TLR10
Numéro d’acquisition du gène Q9BXR5
Alias du gène CD290; MGC104967; MGC126398; MGC126399; TLR10; toll like receptor 10; toll-like receptor 10; UNQ315/PRO358
Symboles de gène TLR10
Espèces hôtes Mouse
Méthode de purification Affinity chromatography
Quantité 100 Tests
Statut réglementaire RUO
Primaire ou secondaire Primary
ID du gène (Entrez) 81793
Espèces cibles Human
Contenu et stockage 4°C, store in dark, DO NOT FREEZE!
Type de produit Antibody
Forme Liquid
Isotype IgG1 κ
Afficher plus de résultats Afficher moins de résultats