Learn More
ACROBiosystems Inc ACROBiosystems Human CD27 Ligand / CD70 Protein, His,Flag Tag (active trimer) (MALS verified)
Numéro de catalogue. 501956444
Encompass
Recombinant Protein;100UG;Human CD27 Ligand, His,Flag Tag (active trimer) (MALS verified) (CDL-H52Da) is expressed from human 293 cells (HEK293). It contains AA Ser 52 - Pro 193 (Accession # P32970-1) trimer Design.;This protein carries a polyhistidine tag at the N-terminus, followed by a flag tag. The protein has a calculated MW of 49.3 kDa. The protein migrates as 50-60 kDa under non-reducing (NR) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD27 Ligand, His,Flag Tag (active trimer) (MALS verified) (Cat. No. CDL-H52Da) at 2 g/mL (100 L/well) can bind Human CD27, Fc Tag (Cat. No. CD7-H5254) with a linear range of 2-125 ng/mL (QC tested).
Numéro de catalogue. 501956444 Fournisseur ACROBiosystems Inc Code fournisseur CDLH52DA100UG
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Recombinant Protein;100UG;Human CD27 Ligand, His,Flag Tag (active trimer) (MALS verified) (CDL-H52Da) is expressed from human 293 cells (HEK293). It contains AA Ser 52 - Pro 193 (Accession # P32970-1) trimer Design.;This protein carries a polyhistidine tag at the N-terminus, followed by a flag tag. The protein has a calculated MW of 49.3 kDa. The protein migrates as 50-60 kDa under non-reducing (NR) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD27 Ligand, His,Flag Tag (active trimer) (MALS verified) (Cat. No. CDL-H52Da) at 2 g/mL (100 L/well) can bind Human CD27, Fc Tag (Cat. No. CD7-H5254) with a linear range of 2-125 ng/mL (QC tested).