Learn More
Invitrogen™ CD195 (CCR5) Monoclonal Antibody (HM-CCR5 (7A4)), PE, eBioscience™, Invitrogen™
Description
Description: CCR5 is the major co-receptor for macrophage-tropic isolates of HIV-1. CCR5, also known as CD195, is a seven transmembrane chemokine receptor capable of binding RANTES, MIP-1 alpha and MIP-1 beta. Knockout mice show a decreased recruitment of macrophages and increased susceptibility to infection. Protein and mRNA has been found in macrophages from thioglycolate-treated and activated T cells. The HM-CCR5(7A4) monoclonal antibody binds the N terminal extracellular domain of mouse CCR5 with no crossreactivity to human CCR5. Applications Reported: This HM-CCR5 (7A4) antibody has been reported for use in flow cytometric analysis. Applications Tested: This HM-CCR5 (7A4) antibody has been tested by flow cytometric analysis of mouse thioglycolate-elicited peritoneal-exudate cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
Spécifications
Spécifications
| Antigène | CD195 (CCR5) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | HM-CCR5 (7A4) |
| Concentration | 0.2 mg/mL |
| Conjugué | PE |
| Formule | PBS with 0.09% sodium azide; pH 7.2 |
| Gène | CCR5 |
| Numéro d’acquisition du gène | P51682 |
| Alias du gène | AM4-7; C-C chemokine receptor type 5; C-C CKR-5; C-C motif chemokine receptor 5 (gene/pseudogene); C-C motif chemokine receptor 5 A159A; CCCKR5; CC-CKR-5; Ccr5; CCR-5; CD195; CD195 antigen; chemokine (C-C motif) receptor 5; chemokine (C-C) receptor 5; chemokine receptor CCR5; chemr13; CKR5; CKR-5; Cmkbr5; HIV-1 fusion coreceptor; IDDM22; MIP-1 alpha receptor |
| Afficher plus de résultats |
For Research Use Only.