Learn More
Invitrogen™ CD138 (Syndecan-1) Monoclonal Antibody (MI15), eFluor™ 450, eBioscience™, Invitrogen™

Description
Description: This MI15 monoclonal antibody reacts with human CD138, also known as syndecan-1. CD138 is a transmembrane protein containing chondroitin sulfate and heparin sulfate moieties responsible for binding to the extracellular matrix components. CD138 is not expressed by mature B cells but is present on pre-B cells and the finally differentiated plasma cells. Applications Reported: This MI15 antibody has been reported for use in flow cytometric analysis. Applications Tested: This MI15 antibody has been pre-diluted and tested by flow cytometric analysis of stimulated normal human peripheral blood cells. This may be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor 450 is an alternative to Pacific Blue. eFluor 450 emits at 446 nm and is excited with the violet laser line (405 nm). Please make sure that your instrument is capable of detecting this fluorochrome. Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser
Spécifications
Spécifications
| Antigène | CD138 (Syndecan-1) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | MI15 |
| Concentration | 5 μL/Test |
| Conjugué | eFluor 450 |
| Formule | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Gène | Sdc1 |
| Numéro d’acquisition du gène | P18827 |
| Alias du gène | AA408134; AA409076; CD antigen 138; CD138; CD138 antigen; heparan sulfate proteoglycan fibroblast growth factor receptor; HSPG; sCD138; SDC; Sdc1; soluble CD138; Sstn; syn-1; Synd; SYND1; Synd-1; SYNDECA; syndecan; syndecan 1; syndecan proteoglycan 1; Syndecan1; syndecan-1; synstatin |
| Afficher plus de résultats |