missing translation for 'onlineSavingsMsg'
Learn More

Catechol, 99+%, Thermo Scientific™

Catalog No. AC158980100
Change view
Click to view available options
10Kg; Fibre drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC158980100 10Kg; Fibre drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC158980100 Supplier Thermo Scientific Chemicals Supplier No. 158980100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 120-80-9
Boiling Point 245.0°C
Flash Point 131°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Fibre drum
Molecular Formula C6H6O2
MDL Number MFCD00002188
Quantity 10kg
Merck Index 15,8111
Synonym pyrocatechol, catechol, 1,2-dihydroxybenzene, 1,2-benzenediol, pyrocatechin, 2-hydroxyphenol, o-benzenediol, pyrocatechine, o-dihydroxybenzene, o-dioxybenzene
Solubility Information Solubility in water: 430g/L (20°C). Other solubilities: soluble in alcohol, benzene, chloroform, ether, and pyridine and aqueous solutions
InChI Key YCIMNLLNPGFGHC-UHFFFAOYSA-N
SMILES OC1=CC=CC=C1O
IUPAC Name benzene-1,2-diol
Molecular Weight (g/mol) 110.11
PubChem CID 289
Viscosity 2.38 mm2/s (110°C)
ChEBI CHEBI:18135
Formula Weight 110.11
Percent Purity 99+%
Chemical Name or Material Catechol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Suspected of causing genetic defects.
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye damage.<b
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cau
EINECSNumber 204-427-5
Recommended Storage Product may darken during storage

RUO – Research Use Only