missing translation for 'onlineSavingsMsg'
Learn More

Catalpol, Thermo Scientific™

Catalog No. AC468942500
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC468942500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC468942500 Supplier Thermo Scientific Chemicals Supplier No. 468942500

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2415-24-9
Molecular Formula C15 H22 O10

Specifications

Quantity 250 mg
Formula Weight 207
Physical Form Powder
Color White to Light-brown
Purity ≥95%
Chemical Name or Material Catalpol
Hazard Category Exclamation mark
EINECSNumber 219-324-0
Recommended Storage Normal conditions