CAS RN 2415-24-9
IUPAC Name:
(2S,3R,4S,5S,6R)-2-{[(1S,2S,4S,5S,6R,10S)-5-hydroxy-2-(hydroxymethyl)-3,9-dioxatricyclo[4.4.0.0²,⁴]dec-7-en-10-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Synonyms:
catalpol catalpinoside
Molecular Weight (g/mol):
362.33
Molecular Formula:
C15H22O10
InChi Key:
LHDWRKICQLTVDL-PZYDOOQISA-N
SMILES:
OC[C@H]1O[C@@H](O[C@@H]2OC=C[C@H]3[C@H](O)[C@@H]4O[C@]4(CO)[C@@H]23)[C@H](O)[C@@H](O)[C@@H]1O