missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Catalase from Bovine Liver
Derived from Bovine liver
$1,032.12
Specifications
| Concentration | ≥3,000 U/mg |
|---|---|
| pH | 7 |
| Content And Storage | -20°C, Protect from moisture |
| Source | Bovine Liver |
| Product Type | Catalase |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAJ1288509
|
N/A | 10 g |
Each for $1,032.12
|
|
|||||
Description
- One unit of enzyme decomposes 1μmol H2O2/min at 25°C at pH 7.0
Specifications
| ≥3,000 U/mg | |
| -20°C, Protect from moisture | |
| Catalase |
| 7 | |
| Bovine Liver | |
| Lyophilized |
Safety and Handling
EINECSNumber : 232-577-1
Recommended Storage : Store at -20°C
RUO – Research Use Only