missing translation for 'onlineSavingsMsg'
Learn More

Catalase from Bovine Liver

Catalog No. AAJ1288509
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ1288509 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ1288509 Supplier Thermo Fisher Scientific Supplier No. J1288509

Derived from Bovine liver

  • One unit of enzyme decomposes 1μmol H2O2/min at 25°C at pH 7.0

Specifications

Concentration ≥3,000 U/mg
pH 7
Content And Storage -20°C, Protect from moisture
Source Bovine Liver
Quantity 10 g
Product Type Catalase
Form Lyophilized

RUO – Research Use Only