missing translation for 'onlineSavingsMsg'
Learn More

Supertek™ Cast Iron Support Ring

Cast iron support ring with side arm and V clamp to fit rods up to 13 mm. The distance from the support clamp center to the center of the ring is 4.5 in. for all rings.

Supplier:  Supertek™ CH12371/3

Encompass

 Afficher plus de versions de ce produit

Numéro de catalogue. S012346


Il en reste null

Description

Description

  • Cast iron support ring
  • With side arm and V clamp to fit rods up to 13 mm
  • The distance from the support clamp center to the center of the ring is 4.5 in.
Spécifications

Identifiants chimiques

Cast Iron
18 mm
Circle
1 Ea.

Spécifications

Cast Iron
3.5 in.
Circle
18 mm
100 mm
1 Ea.
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions