Learn More
Promega Caspase Inhibitors DFS Item
Z-VAD-FMK is a cell-permeant pan caspase inhibitor.
Supplier:  Promega G7231
 View more versions of this product
Catalog No. PR-G7231
Only null left
Explore available promotions
Description
Description
  • A Cell-Permeant Pan Caspase Inhibitor of Apoptosis
  • Irreversibly binds to catalytic site of caspase proteases
  • Provided in DMSO for convenient addition to cell culture or extracts
Note: Z-VAD-FMK is provided at 20mM in DMSO for convenient addition to cell culture or extracts. The peptide is O-methylated in the P1 position on aspartic acid, providing enhanced stability and increased cell permeability. The suggested concentration for use in the anti-Fas mAb-treated Jurkat cell culture model system is 20μM.
Specifications
Specifications
20 mM in DMSO
Irreversibly binds to the catalytic site of Caspase Proteases and can inhibit induction of apoptosis
50 μL
Z-VAD-FMK Caspase
Colorless
-30°C to -10°C
Z-VAD-FMK Caspase Inhibitor
Liquid
Product Suggestions
Product Suggestions