Learn More
Promega Caspase Inhibitors DFS Item
Z-VAD-FMK is a cell-permeant pan caspase inhibitor.
$675.93 - $1126.55
Specifications
Color Colorless
Form Liquid
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Concentration Inhibitors Price Quantity  
Catalog Number Mfr. No. Quantity Concentration Inhibitors Price Quantity  
PR-G7231 DFS Item
View Documents
Promega
G7231
50 μL 20 mM in DMSO Z-VAD-FMK Caspase
Each for $675.93
Only null left
PR-G7232 DFS Item
View Documents
Promega
G7232
125 μL 20 mM in DMSO Z-VAD-FMK Caspase
Each for $1,126.55
Only null left
Description
Description
  • A Cell-Permeant Pan Caspase Inhibitor of Apoptosis
  • Irreversibly binds to catalytic site of caspase proteases
  • Provided in DMSO for convenient addition to cell culture or extracts
Note: Z-VAD-FMK is provided at 20mM in DMSO for convenient addition to cell culture or extracts. The peptide is O-methylated in the P1 position on aspartic acid, providing enhanced stability and increased cell permeability. The suggested concentration for use in the anti-Fas mAb-treated Jurkat cell culture model system is 20μM.
Specifications
Specifications
Colorless
Liquid
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'