Learn More
Invitrogen™ Caspase 11 Monoclonal Antibody (17D9), eBioscience™, Invitrogen™

Description
Description: The monoclonal antibody 17D9 recognizes mouse Caspase-11, a cysteine protease that plays a role in apoptosis and inflammation. Caspase-11 exists as a 43 kDa and 38 kDa pro-caspase form and a 30 and 10 kDa active form. The 17D9 antibody recognizes p43, p38 and p30 forms. Caspase-11 is an upstream protease that activates other caspases, such as caspase-3, -7 and -1, which in turn activate IL-1beta. Under Caspase-11-deficient conditions, cells are unable to process or secrete IL-1beta in response to stimuli and show defects in motility, which are due to disruption of its interaction with Aip1, an actin binding protein. Caspase-11 is homologous to human Caspase-4 (around 60%). Unlike other caspases which tend to be constitutively expressed, caspase-11 is upregulated by LPS treatment in macrophages, lymphocytes and hepatocytes. Applications Reported: This 17D9 antibody has been reported for use in immunoblotting (WB). Applications Tested: This 17D9 antibody has been tested by western blotting at 1-5 μg/mL on J774 cells or stimulated splenocyte lysates. Samples must be reduced for detection. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
Spécifications
Spécifications
| Antigène | Caspase 11 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 17D9 |
| Concentration | 0.5 mg/mL |
| Conjugué | Unconjugated |
| Formule | PBS with 0.09% sodium azide; pH 7.2 |
| Gène | CASP4 |
| Numéro d’acquisition du gène | P70343 |
| Alias du gène | apoptotic cysteine protease Mih1/TX; Casp11; CASP-11; CASP4; CASP-4; caspase 11; caspase 4; caspase 4, apoptosis-related cysteine peptidase; caspase 4, apoptosis-related cysteine protease; Caspase-11; Caspase-4; Caspase-4 subunit 1; Caspase-4 subunit 2; Caspase-4 subunit p10; Caspase-4 subunit p20; Caspl; ICE and Ced-3 homolog 2; ICE(rel)II; ICE(rel)-II; ICEREL-II; ICH2; ICH-2; Ich3; ich-3; Mih1; Mih1/TX; protease ICH-2; Protease ICH-3; Protease TX; TX |
| Afficher plus de résultats |
For Research Use Only.