missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Casein, Vitamin Tested, 30 Mesh

Catalog No. ICN90452080
Change view
Click to view available options
Quantity:
1 lb.
5 lb.
100 lb.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN90452080 100 lb.
ICN90452001 1 lb.
ICN90452005 5 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN90452080 Supplier MP Biomedicals™ Supplier No. 0290452080

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used as cheese, food additive and binder for safety matches.

  • Phosphoprotein
  • Extracted from bovine milk
  • Used as food source
  • CAS: 9000-71-9

Specifications

Product Type Casein
Color Yellow
Format Powder
For Use With (Application) Used as cheese, food additive and binder for safety matches
Quantity 100 lb.
Storage Requirements Store at room temperature (15°C to 30°C)