missing translation for 'onlineSavingsMsg'
Learn More

Casanthranol, MilliporeSigma™ Supelco™

Numéro de catalogue. 111015585
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
111015585 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 111015585 Fournisseur Merck Emd Millipore Code fournisseur C329850G
Il en reste null
Featured Industries: Pharmaceutical (small molecule)

Spécifications

CAS 8024-48-4
Physical Form Neat
MDL Number MFCD00212912
Type Analytical Standard
Quantity 50 g
Grade Analytical Standard