missing translation for 'onlineSavingsMsg'
Learn More

Carvacrol, Thermo Scientific Chemicals

Catalog No. AC368340250
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC368340250 25 g
AC368341000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC368340250 Supplier Thermo Scientific Chemicals Supplier No. 368340250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 499-75-2
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.22
MDL Number MFCD00002236
InChI Key RECUKUPTGUEGMW-UHFFFAOYSA-N
IUPAC Name 2-methyl-5-(propan-2-yl)phenol
SMILES CC(C)C1=CC=C(C)C(O)=C1

Specifications

Melting Point 3.0°C
Density 0.9760 g/mL
Boiling Point 238.0°C
Flash Point 106°C
Linear Formula (CH3)2CHC6H3(CH3)OH
Quantity 25 g
Merck Index 15, 1872
Specific Gravity 0.976
Solubility Information Solubility aq. soln.: insoluble. Other solubilities: soluble in ethanol and ether
Viscosity 28.5 mPa.s (20°C)
Formula Weight 150.22
Chemical Name or Material Carvacrol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Causes skin irritation.

Causes serious eye irritation.

May cause an allergic skin reaction.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Wear protective gloves/protective clothing/eye protection/face protection.

Call a POISON CENTER or doctor/physician if you feel unwell.

IF ON SKIN: Wash with plenty of soap and water.

If skin irritation or rash occurs: Get medical advice/attention.

If eye irritation persists: Get medical advice/attention.

EINECSNumber 207-889-6

RUO – Research Use Only