missing translation for 'onlineSavingsMsg'
Learn More

Carvacrol 98.0+%, TCI America™

Catalog No. C002625G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C002625G 25 g
C0026100G 100 g
C0026500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C002625G Supplier TCI America Supplier No. C002625G

Chemical Identifiers

CAS 499-75-2
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.22
MDL Number MFCD00002236
InChI Key RECUKUPTGUEGMW-UHFFFAOYSA-N
Synonym carvacrol, 5-isopropyl-2-methylphenol, isopropyl-o-cresol, o-thymol, antioxine, isothymol, karvakrol, 5-isopropyl-o-cresol, 2-p-cymenol, 2-hydroxy-p-cymene
PubChem CID 10364
ChEBI CHEBI:3440
IUPAC Name 2-methyl-5-(propan-2-yl)phenol
SMILES CC(C)C1=CC=C(C)C(O)=C1

Specifications

Color Yellow
Boiling Point 238°C
Quantity 25 g
Formula Weight 150.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Carvacrol
EINECSNumber (4)-0057
RTECSNumber FI1225000
TSCA Yes