missing translation for 'onlineSavingsMsg'
Learn More

Carminic acid, ≥96% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111012852
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012852 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012852 Supplier Merck Emd Millipore Supplier No. 1129825MG

Carminic acid is an anthraquinone red dye. It is majorly used in textile industry as pigmenting agent.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 1260-17-9
Physical Form Neat
Linear Formula C22H20O13
MDL Number MFCD00167028
Synonym Natural Red 4
Type Analytical Standard
Quantity 25 mg
Formula Weight 492.39
Percent Purity ≥96% (HPLC)
Grade Analytical Standard