missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cardiotrophin 1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103114
Description
Antibody detects endogenous levels of total Cardiotrophin-1.
Cardiotrophin-1 (CT-1) is a member of the IL-6 family of cytokines, which signal through gp130 receptor complexes. gp130 complexes with several different receptor subunits to transmit signals from Cardiotrophin-1, IL-6, LIF, OSM, CNTF and IL-11. Cardiotrophin-1 binds to and activates the leukemia inhibitory factor (LIF) receptor/gp130 receptor complex and has been shown to induce hypertrophy in cardiac myocytes in vitro. Cardiotrophin-1, a secreted protein expressed at high levels in myocardium during cardiogenesis, has been shown to promote proliferation and survival of embryonic cardiomyocytes, suggesting a role for Cardiotrophin-1 in the activation of gp130 during cardiac development.Identifiants chimiques
| Cardiotrophin 1 | |
| -20°C | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| CTF1 | |
| cardiophin 1; Cardiotrophin; cardiotrophin 1; Cardiotrophin1; cardiotrophin-1; CT 1; CT1; CT-1; CT-1CT1; CTF 1; Ctf1; IL-6 cytokine family | |
| CTF1 | |
| Primary | |
| Affinity Chromatography |
| 1 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q16619, Q60753, Q63086 | |
| 13019, 1489, 29201 | |
| A synthesized peptide derived from human CTF1(Accession Q16619), corresponding to amino acid residues L33-V83. | |
| Antibody |
Spécifications
| Cardiotrophin 1 | |
| Polyclonal | |
| Unconjugated | |
| CTF1 | |
| cardiophin 1; Cardiotrophin; cardiotrophin 1; Cardiotrophin1; cardiotrophin-1; CT 1; CT1; CT-1; CT-1CT1; CTF 1; Ctf1; IL-6 cytokine family | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 13019, 1489, 29201 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q16619, Q60753, Q63086 | |
| CTF1 | |
| A synthesized peptide derived from human CTF1(Accession Q16619), corresponding to amino acid residues L33-V83. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |