missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carboxymethyl Cellulose Sodium Salt, Medium Viscosity, MP Biomedicals
Chemical Identifiers
| CAS | 9004-32-4 |
|---|---|
| Molecular Formula | (C12 H14 O9 R6)n |
| Molecular Weight (g/mol) | 263.20 |
| MDL Number | MFCD00081472 |
| InChI Key | DPXJVFZANSGRMM-UHFFFAOYNA-N |
| Synonym | carboxymethylcellulose sodium usp, celluvisc tn, carmellose sodium jp17, sodium dextrose acetate, c.m.c. tn |
| PubChem CID | 23706213 |
| SMILES | [Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
ICN15056090
|
N/A | 500 g |
N/A
|
|
|||||
Chemical Identifiers
| (C12 H14 O9 R6)n | |
| MFCD00081472 | |
| carboxymethylcellulose sodium usp, celluvisc tn, carmellose sodium jp17, sodium dextrose acetate, c.m.c. tn | |
| [Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O |
Specifications
| 9004-32-4 | |
| (C12 H14 O9 R6)n | |
| carboxymethylcellulose sodium usp, celluvisc tn, carmellose sodium jp17, sodium dextrose acetate, c.m.c. tn | |
| [Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O | |
| 263.20 | |
| Powder |
| White | |
| MFCD00081472 | |
| DPXJVFZANSGRMM-UHFFFAOYNA-N | |
| sodium;2,3,4,5,6-pentahydroxyhexanal;acetate | |
| 23706213 | |
| Carboxymethyl cellulose sodium salt |
Safety and Handling
Recommended Storage : Store at Room Temperature (15 to 30°C).