missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Carboxymethyl Cellulose Sodium Salt, Medium Viscosity, MP Biomedicals

Manufacturer:  Mp Biomedicals 0215056090

 View more versions of this product

Catalog No. ICN15056090



Specifications

Specifications

Carboxymethyl cellulose sodium salt
500g
C8H15NaO8
c.m.c. tn, carboxymethylcellulose sodium usp, carmellose sodium jp17, celluvisc tn, sodium dextrose acetate
CC(=O)[O-].C(C(C(C(C(C=O)O)O)O)O)O.[Na+]
262.19
Powder
White
9004-32-4
MFCD00081472
QMGYPNKICQJHLN-UHFFFAOYSA-M
sodium;2,3,4,5,6-pentahydroxyhexanal;acetate
23706213

Chemical Identifiers

9004-32-4
262.19
QMGYPNKICQJHLN-UHFFFAOYSA-M
23706213
CC(=O)[O-].C(C(C(C(C(C=O)O)O)O)O)O.[Na+]
C8H15NaO8
MFCD00081472
c.m.c. tn, carboxymethylcellulose sodium usp, carmellose sodium jp17, celluvisc tn, sodium dextrose acetate
sodium;2,3,4,5,6-pentahydroxyhexanal;acetate
Safety and Handling

Safety and Handling

Recommended Storage : Store at Room Temperature (15 to 30°C).

Documents